Benzamidine hydrochloride hydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20122.0002 8.20122.0025 8.20122.1000
CA8.20122.0002CS 6968 CAD
CA8.20122.0002 CA8.20122.0025 CA8.20122.1000
Benzamidine hydrochloride hydrate for synthesis, Sigma-Aldrich®
Benzamidine hydrochloride hydrate
Formula: C₆H₅C(=NH)NH₂·HCl·H₂O
MDL Number: MFCD00066285
CAS Number: 206752-36-5

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 80 - 84 °C
pH 5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR