Dimethyl acetal for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Acetaldehyde dimethyl acetal, 1,1-Dimethoxyethane

M-Clarity™ quality level = MQ200

8.20002.0025 8.20002.0100 8.20002.9010
CA8.20002.0025CS 9133 CAD
CA8.20002.0025 CA8.20002.0100 CA8.20002.9010
Dimethyl acetal for synthesis, Sigma-Aldrich®
Dimethyl acetal
Formula: C₄H₁₀O₂
Boiling Pt: 64 °C (1013 hPa)
Melting Pt: –113 °C
Density: 0.8505 g/cm³ (20 °C)
MDL Number: MFCD00008493
CAS Number: 534-15-6
EINECS: 208-589-8
UN: 2377
ADR: 3,II
Merck Index: 13,03253

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -113 °C
Molar Mass 90.12 g/mol
Boiling Point 63 - 65 °C (1013 hPa)
Vapor Pressure 200 hPa (20 °C)
Flash Point -17 °C
Refractive Index 1.3665 (20 °C, 589 nm)
Density 0.85 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR