Tetrabutylammonium hydrogen sulfate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18858.0100 8.18858.0500 8.18858.9010
CA8.18858.0100CS 5426 CAD
CA8.18858.0100 CA8.18858.0500 CA8.18858.9010
Tetrabutylammonium hydrogen sulfate for synthesis, Sigma-Aldrich®
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
MDL Number: MFCD00011637
CAS Number: 32503-27-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 168 - 174 °C
Molar Mass 339.54 g/mol
Bulk Density 650 kg/m3
pH 1 - 2 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR