2,6-Dimethyl-4-heptanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18831.0100 8.18831.1000
CA8.18831.0100CS 804 CAD
CA8.18831.0100 CA8.18831.1000
2,6-Dimethyl-4-heptanone for synthesis, Sigma-Aldrich®
2,6-Dimethyl-4-heptanone
Formula: C₉H₁₈O
MDL Number: MFCD00008940
CAS Number: 108-83-8
UN: 1157
ADR: 3,III

Order Now

Specification Test Results

Solubility 0.5 g/l (20 °C)
Melting Point -41 °C
Molar Mass 142.24 g/mol
Boiling Point 163 - 173 °C (1013 hPa)
Vapor Pressure 1.6 hPa (20 °C)
Flash Point 47 °C
Refractive Index 1.4143 (20 °C, 589 nm)
Explosion Limit 0.8 - 6.2 %(V)
Density 0.81 g/cm3 (20 °C)
Ignition Point 345 °C
Saturation concentration 9 g/m3 (20 °C) Air
Evaporation number 48
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR