Triethylsilane zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


MQ200

8.18806.0010 8.18806.0050
CA8.18806.0010CS 236145 CAD
CA8.18806.0010 CA8.18806.0050
Triethylsilane zS, Sigma-Aldrich®
Triethylsilane
Formula: C₆H₁₆Si
MW: 116.28 g/mol
Boiling Pt: 107
Melting Pt: -156.9
Flash Pt: -3
MDL Number: MFCD00009018
CAS Number: 617-86-7
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 116.28 g/mol
Boiling Point 107 - 109 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point -6 °C
Refractive Index 1.4117 (20 °C, 589 nm)
Density 0.73 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR