p-Toluenesulfonic acid monohydrate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate, 4-Methylbenzenesulfonic acid hydrate, Toluene-4-sulfonic acid monohydrate
M-Clarity™ quality level = MQ200
8.14725.0100
8.14725.1000
8.14725.5000
CA8.14725.0100CS
5287
CAD
CA8.14725.0100
CA8.14725.1000
CA8.14725.5000
p-Toluenesulfonic acid monohydrate for synthesis, Sigma-Aldrich®
p-Toluenesulphonic acid monohydrate
Formula:
C₇H₈O₃S·1H₂O MW: 190.22 g/mol Flash Pt: 180 |
MDL Number:
MFCD00142137 CAS Number: 6192-52-5 UN: 2585 ADR: 8 III Merck Index: 12,09671 |
Specification Test Results
Assay (acidimetric, calc. on anhydrous substance) | ≥ 98.0 % |
Identity (IR) | passes test |
Water (K. F.) | 8.0 - 12.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...