Glycolic acid ≥70% in water, special quality, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Hydroxy acid, 2-Hydroxyacetic acid

M-Clarity™ quality level = MQ200

8.14662.1000 8.14662.5000 8.14662.9026
CA8.14662.1000CS 4351 CAD
CA8.14662.1000 CA8.14662.5000 CA8.14662.9026
Glycolic acid ≥70% in water, special quality, Sigma-Aldrich®
Glycolic acid
Formula: C₂H₄O₃
MW: 76.05 g/mol
Boiling Pt: 266
MDL Number: MFCD00004312
CAS Number: 79-14-1
UN: 3265
ADR: 8 II
Merck Index: 13,04511

Order Now

Specification Test Results

Assay (acidimetric) 69.0 - 74.0 %
Color number (Hazen) ≤ 30
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Density (d 20\u2007°C/ 4\u2007°C) 1.260 - 1.280
Heavy metals (as Pb) ≤ 3 ppm
pH-value 0.0 - 1.0
Refractive index (n 20°/D) 1.410 - 1.415


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR