O-Phenylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14538.0005 8.14538.0250 8.14538.1000
CA8.14538.0005CS 64655 CAD
CA8.14538.0005 CA8.14538.0250 CA8.14538.1000
O-Phenylenediamine for synthesis, Sigma-Aldrich®
o-Phenylenediamine
Formula: C₆H₈N₂
MDL Number: MFCD00007721
CAS Number: 95-54-5
UN: 1673
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 54 g/l (20 °C)
Melting Point 99 - 102 °C
Molar Mass 108.14 g/mol
Bulk Density 530 kg/m3
Boiling Point 257 °C (1013 hPa)
Vapor Pressure 0.013 hPa (20 °C)
Flash Point 110 °C
Explosion Limit 1.5 %(V)
Density 1.14 g/cm3 (20 °C)
pH 7 - 8 (50 g/l, H2O, 20 °C)
Ignition Point 540 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR