Sodium bromate 99,0 Iodometry zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


MQ200

8.14368.0250 8.14368.1000 8.14368.9025
CA8.14368.0250CS 16911 CAD
CA8.14368.0250 CA8.14368.1000 CA8.14368.9025
Sodium bromate 99,0 Iodometry zS, Sigma-Aldrich®
Sodium bromate
Formula: BrNaO₃
MW: 150.89 g/mol
Boiling Pt: 1390
Melting Pt: 381
MDL Number: MFCD00003476
CAS Number: 7789-38-0
UN: 1494
ADR: 5.1 II
Merck Index: 12,08736

Order Now

Specification Test Results

Solubility 364 g/l (20 °C)
Melting Point 381 °C (decomposition)
Molar Mass 150.89 g/mol
Density 3.3 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral,Aqueous solution
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR