Sodium dehydracetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Dehydracetic acid sodium salt, Dehydroacetic acid sodium salt, Sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide, Sodium dehydroacetate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate

M-Clarity™ quality level = MQ200

8.14085.0005 8.14085.0250 8.14085.1000
CA8.14085.0005CS 62604 CAD
CA8.14085.0005 CA8.14085.0250 CA8.14085.1000
Sodium dehydracetate for synthesis, Sigma-Aldrich®
Sodium dehydracetate
Formula: C₈H₇NaO₄
MDL Number: MFCD00040583
CAS Number: 4418-26-2

Order Now

Specification Test Results

Assay(HClO4) ≥98.0%
Identity( IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR