Hexamethyldisiloxane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Silicone fluid, HMDSO

M-Clarity™ quality level = MQ200

8.14051.0100 8.14051.0500
CA8.14051.0100CS 1578 CAD
CA8.14051.0100 CA8.14051.0500
Hexamethyldisiloxane for synthesis, Sigma-Aldrich®
Hexamethyldisiloxane
Formula: C₆H₁₈OSi₂
MW: 162.38 g/mol
Boiling Pt: 101
Melting Pt: -59
Flash Pt: 0.6
MDL Number: MFCD00008265
CAS Number: 107-46-0
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03d0d18c44dfa536ffcc853891b6f469333a38c5&vid=4959fefab2528efb6003e1f23c57966b65984db6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Melting Point -68 °C
Molar Mass 162.38 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 20 hPa (20 °C)
Flash Point 4 °C
Refractive Index 1.3771 (20 °C, 589 nm)
Explosion Limit 0.5 - 21.8 %(V)
Density 0.76 g/cm3 (25 °C)
Ignition Point 340 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR