Diethyl carbonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.02898.0100 8.02898.0500 8.02898.1000
CA8.02898.0100CS 2035 CAD
CA8.02898.0100 CA8.02898.0500 CA8.02898.1000
Diethyl carbonate for synthesis, Sigma-Aldrich®
Diethyl carbonate
Formula: C₅H₁₀O₃
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0.9751 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009107
CAS Number: 105-58-8
EINECS: 203-311-1
UN: 2366
ADR: 3,III
Merck Index: 13,03815

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -43 °C
Molar Mass 118.13 g/mol
Boiling Point 126 °C (1013 hPa)
Vapor Pressure 11 hPa (20 °C)
Flash Point 33 °C
Refractive Index 1.384 (20 °C)
Explosion Limit 1.4 - 11.0 %(V)
Density 0.97 g/cm3 (20 °C)
pH 6.3 (1 g/l, H2O)
Ignition Point 445 °C
Evaporation number 14
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR