Cysteaminium chloride zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Aminoethanethiol hydrochloride, Cysteamine Hydrochloride, 2-Mercaptoethylamine hydrochloride, 2-MEA, β-Mercaptoethylamine hydrochloride, Mercaptoethylamine-HCl, H-Cys-NH2·HCl

MQ200

8.02835.0010 8.02835.0025 8.02835.0100
CA8.02835.0010CS 10624 CAD
CA8.02835.0010 CA8.02835.0025 CA8.02835.0100
Cysteaminium chloride zS, Sigma-Aldrich®
Cysteaminium chloride
Formula: C₂H₇NS·HCl
MW: 113.61 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012904
CAS Number: 156-57-0
EINECS: 205-858-1

Order Now

Specification Test Results

Assay (iodometric) ≥ 97.0 %
Melting range (lower value) ≥ 61 °C
Melting range (upper value) ≤ 67 °C
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR