(±)-1,3-Butanediol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.01964.1000 8.01964.9025 8.01964.9200
CA8.01964.1000CS 1505 CAD
CA8.01964.1000 CA8.01964.9025 CA8.01964.9200
(±)-1,3-Butanediol for synthesis, Sigma-Aldrich®
(±)-1,3-Butanediol
Formula: C₄H₁₀O₂
Boiling Pt: 206…208 °C (1013 hPa)
Melting Pt: Max. -50 °C
Density: 1.004…1.006 g/cm³ (20 °C)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00004554
CAS Number: 107-88-0
EINECS: 203-529-7
Merck Index: 13,01566

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 90.12 g/mol
Boiling Point 207.5 °C (1.013 hPa)
Vapor Pressure 0.08 hPa (20 °C)
Flash Point 121 °C
Refractive Index 1.4395 (20 °C, 589 nm)
Explosion Limit 1.8 - 9.4 %(V)
Density 1.00 g/cm3 (20 °C)
pH 6 - 7 (1 g/l, H2O)
Ignition Point 440 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR