Chlorobenzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

M-Clarity™ quality level = MQ200

8.01791.9025 8.01791.0100 8.01791.1000 8.01791.2500
CA8.01791.9025EA 773 CAD
CA8.01791.9025 CA8.01791.0100 CA8.01791.1000 CA8.01791.2500
Chlorobenzene for synthesis, Sigma-Aldrich®
Chlorobenzene
Formula: C₆H₅Cl
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.4 g/l (20 °C)
Melting Point -45 °C
Molar Mass 112.56 g/mol
Boiling Point 132 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Flash Point 27 °C
Refractive Index 1.5248 (20 °C, 589 nm)
Explosion Limit 1.3 - 11 %(V)
Density 1.11 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Saturation concentration 54 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR