Diisopropyl ether stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Isopropyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
M-Clarity™ quality level = MQ200

8.00866.1000 8.00866.2500 8.00866.9040 8.00866.9143
CA8.00866.1000CS 781 CAD
CA8.00866.1000 CA8.00866.2500 CA8.00866.9040 CA8.00866.9143
Diisopropyl ether stabilised for synthesis, Sigma-Aldrich®
Diisopropyl ether
Formula: (CH₃)₂CHOCH(CH₃)₂
Boiling Pt: 68.5 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.726 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3,II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 12 g/l (20 °C)
Melting Point -86 °C
Molar Mass 102.18 g/mol
Boiling Point 67 - 70 °C (1013 hPa)
Vapor Pressure 175 hPa (20 °C)
Flash Point -22 °C
Refractive Index 1.3679 (20 °C, 589 nm)
Explosion Limit 1.0 - 21 %(V)
Density 0.72 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 405 °C
Saturation concentration 751 g/m3 (20 °C) Air
Evaporation number 1.6
Water absorption 60 g/kg (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR