Methyl acrylate stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.00841.0100 8.00841.1000
CA8.00841.0100CS 1934 CAD
CA8.00841.0100 CA8.00841.1000
Methyl acrylate stabilized for synthesis, Sigma-Aldrich®
Methyl acrylate (monomer)
Formula: C₄H₆O₂
MDL Number: MFCD00008627
CAS Number: 96-33-3
EINECS: 202-500-6
UN: 1919
ADR: 3,II

Order Now

Specification Test Results

Solubility 52 g/l (25 °C)
Melting Point -74 °C
Molar Mass 86.09 g/mol
Boiling Point 79 - 81 °C (1013 hPa)
Vapor Pressure 93 hPa (20 °C)
Flash Point -2.7 °C
Refractive Index 1.4021 (20 °C, 589 nm)
Explosion Limit 2.8 - 18.6 %(V)
Density 0.96 g/cm3 (20 °C)
Ignition Point 415 °C
Water absorption 250 g/kg (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR