1,2-Ethanedithiol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1.2-Ethanedithiol, 1,2-Dimercaptoethane

M-Clarity™ quality level = MQ200

8.00795.0100 8.00795.0500
CA8.00795.0100CS 11389 CAD
CA8.00795.0100 CA8.00795.0500
1,2-Ethanedithiol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
1,2-Ethanedithiol
Formula: C₂H₆S₂
MW: 94.2 g/mol
Boiling Pt: 146…149 °C (1013 hPa)
Melting Pt: -41
Density: 1.374 g/cm³ (20 °C)
Flash Pt: 44
Storage Temperature: Ambient
MDL Number: MFCD00004892
CAS Number: 540-63-6
EINECS: 208-752-3
UN: 1228
ADR: 3 (6.1) III
Merck Index: 13,03760

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -41 °C
Molar Mass 94.20 g/mol
Boiling Point 146 - 149 °C
Vapor Pressure 5.3 hPa (23 °C)
Flash Point 44 °C
Density 1.12 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR