Succinic anhydride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Dihydrofuran-2,5-dione
M-Clarity™ quality level = MQ200
8.00683.0100
8.00683.0500
8.00683.1000
8.00683.9050
CA8.00683.0100CS
1757
CAD
CA8.00683.0100
CA8.00683.0500
CA8.00683.1000
CA8.00683.9050
Succinic anhydride for synthesis, Sigma-Aldrich®
Succinic anhydride
Formula:
C₄H₄O₃ MW: 100.07 g/mol Boiling Pt: 261 °C (1013 hPa) Melting Pt: 120 °C Density: 1.503 g/cm³ (20 °C) Flash Pt: 157 °C |
MDL Number:
MFCD00005525 CAS Number: 108-30-5 EINECS: 203-570-0 UN: 3261 ADR: 8,II Merck Index: 12,09039 |
Specification Test Results
Solubility | 67 g/l (20 °C) Hydrolysis |
Melting Point | 116 - 120 °C |
Molar Mass | 100.07 g/mol |
Bulk Density | 550 - 600 kg/m3 |
Boiling Point | 261 °C (1013 hPa) |
Vapor Pressure | 1.3 hPa (92 °C) |
Flash Point | 157 °C |
Density | 1.48 g/cm3 (20 °C) |
pH | 2.7 (20 g/l, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...