Benzenesulfonyl chloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.00470.0100
8.00470.0500
8.00470.2500
CA8.00470.0100CS
1780
CAD
CA8.00470.0100
CA8.00470.0500
CA8.00470.2500
Benzenesulfonyl chloride for synthesis, Sigma-Aldrich®
Benzenesulphonyl chloride
Formula:
C₆H₅ClO₂S Boiling Pt: 251 °C (1013 hPa) Melting Pt: 15 °C Density: 1.3842 g/cm³ (20 °C) Flash Pt: 119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007426 CAS Number: 98-09-9 EINECS: 202-636-6 UN: 2225 ADR: 8,III Merck Index: 13,01072 |
Specification Test Results
Solubility | (20 °C) insoluble,(decomposition) |
Melting Point | 15 °C |
Molar Mass | 176.62 g/mol |
Boiling Point | 252 °C (1013 hPa) decomposes |
Vapor Pressure | 6 hPa (20 °C) |
Flash Point | 123 °C |
Refractive Index | 1.553 (20 °C, 589 nm) |
Density | 1.38 g/cm3 (20 °C) |
Ignition Point | 450 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...