Benzenesulfonic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.00468.0005
8.00468.0250
CA8.00468.0005CS
92688
CAD
CA8.00468.0005
CA8.00468.0250
Benzenesulfonic acid for synthesis, Sigma-Aldrich®
Benzenesulphonic acid
Formula:
C₆H₆O₃S Boiling Pt: 319 °C (1013 hPa) Melting Pt: 50 °C Density: 1.32 g/cm³ (47 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011689 CAS Number: 98-11-3 EINECS: 202-638-7 UN: 2585 ADR: 8,III Merck Index: 13,01070 |
Specification Test Results
Solubility | 930 g/l (25 °C) |
Melting Point | 43 °C |
Molar Mass | 158.17 g/mol |
Boiling Point | 171 °C (0.13 hPa) |
Vapor Pressure | (20 °C) |
Flash Point |
|
Density | 1.32 g/cm3 (47 °C) |
pH | 2 (H2O, 20 °C) (saturated solution) |
Hazard Class | 8 |
Hazard Class Description | Corrosive material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...