N,O-Bis(trimethylsilyl)trifluoroacetamide for gas chromatography, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

M-Clarity™ quality level = MQ100

1.10255.0005 1.10255.0025
CA1.10255.0005CS 49428 CAD
CA1.10255.0005 CA1.10255.0025
N,O-Bis(trimethylsilyl)trifluoroacetamide for gas chromatography, Supelco®
N,O-Bis(trimethylsilyl)trifluoroacetamide
Formula: C₈H₁₈F₃NOSi₂
MW: 257.4 g/mol
Boiling Pt: 145
Melting Pt: -41
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 34
MDL Number: MFCD00008269
CAS Number: 25561-30-2
EINECS: 247-103-9
UN: 1993
ADR: 3

Order Now

Specification Test Results

Solubility (20 °C) Hydrolysis
Molar Mass 257.40 g/mol
Boiling Point 145 °C (1013 hPa)
Flash Point 34 °C
Density 0.97 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR