o-Xylene reference substance for gas chromatography, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1,2-Dimethylbenzene
1.09798.0005
1.09798.0005
CA1.09798.0005CS
153958.36
CAD
EM1.09798.0005
CA1.09798.0005
o-Xylene reference substance for gas chromatography, Supelco®
o-Xylene
Formula:
C₈H₁₀ MW: 106.17 g/mol Boiling Pt: 144 °C (1013 hPa) Melting Pt: –25 °C Density: 0.88 g/cm³ (20 °C) Flash Pt: 30 °C (closed cup) |
MDL Number:
MFCD00008519 CAS Number: 95-47-6 UN: 1307 ADR: 3, Merck Index: 12,10214 |
Specification Test Results
Solubility | 0.18 g/l (20 °C) |
Melting Point | -25 °C |
Molar Mass | 106.17 g/mol |
Boiling Point | 144.4 °C (1013 hPa) |
Vapor Pressure | 7 hPa (20 °C) |
Flash Point | 30 °C |
Explosion Limit | 1.0 - 7.6 %(V) |
Density | 0.88 g/cm3 (20 °C) |
Ignition Point | 465 °C |
Saturation concentration | 29 g/m3 (20 °C) Air |
Evaporation number | 13.5 |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...