n-Tetradecane reference substance for gas chromatography, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Tetradecane
1.09608.0005
1.09608.0005
CA1.09608.0005CS
240094
CAD
EM1.09608.0005
CA1.09608.0005
n-Tetradecane reference substance for gas chromatography, Supelco®
n-Tetradecane
Formula:
H₃C(CH₂)₁₂CH₃ MW: 198.39 g/mol Boiling Pt: 253.7 °C (1013 hPa) Melting Pt: 6 °C Density: 0.7628 g/cm³ (20 °C) Flash Pt: 102 °C (closed cup) |
MDL Number:
MFCD00008986 CAS Number: 629-59-4 |
Specification Test Results
Solubility |
|
Melting Point | 6 °C |
Molar Mass | 198.39 g/mol |
Boiling Point | 254 °C |
Vapor Pressure | 1.55 Pa (25 °C) |
Flash Point | 102 °C |
Explosion Limit | 0.5 - 6.5 %(V) |
Density | 0.76 g/cm3 (20 °C) |
Ignition Point | 202 °C |
Viscosity kinematic | 3 mm2/s (40 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...