n-Hexadecane reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

1.09605.0005
CA1.09605.0005CS 154469 CAD
CA1.09605.0005
n-Hexadecane reference substance for gas chromatography, Supelco®
n-Hexadecane
Formula: H₃C(CH₂)₁₄CH₃
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.77331 g/cm³ (20 °C)
Flash Pt: 135 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 18 °C
Molar Mass 226.44 g/mol
Boiling Point 287 °C
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 135 °C
Density 0.77 g/cm3 (20 °C)
Viscosity kinematic  - 7 mm2/s (40 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR