Folin-Ciocalteu's reagent for spectrophotometry, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Folin & Ciocalteu’s phenol reagent
1.09001.0100
1.09001.0500
CA1.09001.0100CS
962
CAD
CA1.09001.0100
CA1.09001.0500
Folin-Ciocalteu's reagent for spectrophotometry, Supelco®
Folin-Ciocalteu's reagent
Density:
1.22 g/cm³ (20 °C) |
MDL Number:
MFCD00132625 UN: 3264 ADR: 8,III |
Specification Test Results
Solubility | (20 °C) soluble |
Density | 1.24 g/cm3 (20 °C) |
pH |
|
Hazard Class | 8 |
Hazard Class Description | Corrosive material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...