Thiourea, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Sulfourea, Thiocarbamide, Sulphourea

1.07979.0250 1.07979.1000
CA1.07979.0250CS 4711 CAD
CA1.07979.0250 CA1.07979.1000
Thiourea, GR for analysis, Supelco®
Thiourea
Formula: NH₂CSNH₂
MW: 76.12 g/mol
Melting Pt: 174…177 °C
Density: 1.435 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9 III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 137 g/l (20 °C)
Melting Point 169 - 173 °C
Molar Mass 76.12 g/mol
Bulk Density 640 kg/m3
Vapor Pressure (20 °C)
Density 1.405 g/cm3 (20 °C)
pH 6 - 8 (50 g/l, H2O, 20 °C)
Ignition Point 440 °C Dust
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR