1,10-Phenanthroline hydrochloride monohydrate ≥99.5% (by anhydrous basis), GR Reag. Ph. Eur. for analysis, redox indicator, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate
1.07223.0010
1.07223.0100
CA1.07223.0010CS
44798
CAD
CA1.07223.0010
CA1.07223.0100
1,10-Phenanthroline hydrochloride monohydrate ≥99.5% (by anhydrous basis), GR Reag. Ph. Eur. for analysis, redox indicator, Supelco®
1,10-Phenanthroline hydrochloride monohydrate
- reagent for iron(II)
Formula:
C₁₂H₉ClN₂·1H₂O MW: 234.68 g/mol Melting Pt: 224…226 °C Density: 1.385 g/cm³ (–100.15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150061 CAS Number: 18851-33-7 EINECS: 223-325-1 UN: 2811 ADR: 6.1 I |
Specification Test Results
Assay (alcalimetric, calculated on anhydrous substance) | ≥ 99.5 % |
Identity (IR-spectrum) | passes test |
Melting point | about 215 °C, with decomposition |
Substances insoluble in water | passes test |
Sulfated ash | ≤ 0.2 % |
Water (according to Karl Fischer) | 7.0 - 8.5 % |
Suitability as indicator | passes test |
Sensitivity test (at Iron II ions) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...