Ninhydrin, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.06762.0010 1.06762.0100 1.06762.1000
CA1.06762.0010CS 20772 CAD
CA1.06762.0010 CA1.06762.0100 CA1.06762.1000
Ninhydrin, GR for analysis, Supelco®
Ninhydrin
Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250…258 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 20 g/l (20 °C)
Melting Point 250 - 258 °C (decomposition)
Molar Mass 178.15 g/mol
Bulk Density 680 kg/m3
pH 4.6 - 5.0 (10 g/l, H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR