Potassium carbonate - sodium carbonate, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.06683.0500 1.06683.2500
CA1.06683.0500CS 2790 CAD
CA1.06683.0500 CA1.06683.2500
Potassium carbonate - sodium carbonate, GR for analysis, Supelco®
Potassium carbonate - sodium carbonate
Formula: CKNaO₃
Density: 2.47 g/cm³ (20 °C)
MDL Number: MFCD00053407
CAS Number: 10424-09-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Bulk Density 1750 kg/m3
Density 2.47 g/cm3 (20 °C)
pH 11.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR