Sodium nitroprusside dihydrate, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06541.0025 1.06541.0100 1.06541.0500
CA1.06541.0025CS 39257 CAD
CA1.06541.0025 CA1.06541.0100 CA1.06541.0500
Sodium nitroprusside dihydrate, GR for analysis, Supelco®
Sodium nitroprusside dihydrate

EMD # 1.06541.0100 replaces and is identical to EMD # SX0675-2.

Formula: Na₂[Fe(CN)₅NO]·2H₂O
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149192
CAS Number: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,II
Merck Index: 13,08722

Order Now

Specification Test Results

Solubility 400 g/l (20 °C) (slow decomposition)
Molar Mass 297.95 g/mol
Bulk Density 1000 kg/m3
Density 1.71 g/cm3 (20 °C)
pH 5 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR