1-Naphthol, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06223.0050 1.06223.0250
CA1.06223.0050CS 23423 CAD
CA1.06223.0050 CA1.06223.0250
1-Naphthol, GR for analysis, Supelco®
1-Naphthol
Formula: C₁₀H₈O
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.224 g/cm³ (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003930
CAS Number: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,06409

Order Now

Specification Test Results

Solubility 0.1 g/l (20 °C)
Melting Point 94 - 96 °C
Molar Mass 144.17 g/mol
Bulk Density 450 kg/m3
Boiling Point 288 °C (1013 hPa)
Vapor Pressure 1.3 hPa (94 °C)
Flash Point 125 °C
Density 1.28 g/cm3 (20 °C)
Ignition Point 510 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR