2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

M-Clarity™ quality level = MQ100

1.15440.1000 1.15440.2500
CA1.15440.1000CS 2191 CAD
CA1.15440.1000 CA1.15440.2500
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
2,2,4-Trimethylpentane

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2
Melting Pt: -107
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.56 mg/l (25 °C)
Melting Point -107 °C
Molar Mass 114.23 g/mol
Boiling Point 99 °C (1013 hPa)
Vapor Pressure 51 hPa (20 °C)
Flash Point -14 °C
Explosion Limit 1 - 5.5 %(V)
Density 0.69 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 410 °C
Saturation concentration 239 g/m3 (20 °C)
Evaporation number 2.3
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR