Acetonitrile for DNA synthesis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
1.13212.2500
1.13212.4000
CA1.13212.2500CS
5596
CAD
CA1.13212.2500
CA1.13212.4000
Acetonitrile for DNA synthesis, Supelco®
Acetonitrile
Formula:
H₃CCN Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –45.7 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3, REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -45.7 °C |
Molar Mass | 41.05 g/mol |
Boiling Point | 81.6 °C (1013 hPa) |
Vapor Pressure | 97 hPa (20 °C) |
Flash Point | 2 °C |
Refractive Index | 1.34 |
Explosion Limit | 3.0 - 17 %(V) |
Density | 0.786 g/cm3 (20 °C) |
pH | (H2O) no data available |
Ignition Point | 524 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...