Dichloromethane, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Methylene chloride

1.06048.0500 1.06048.2500
CA1.06048.0500CS 1713 CAD
CA1.06048.0500 CA1.06048.2500
Dichloromethane, Uvasol® for spectroscopy, Supelco®
Dichloromethane

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 20 g/l (20 °C)
Melting Point -95 °C
Molar Mass 84.93 g/mol
Boiling Point 40 °C (1013 hPa)
Vapor Pressure 475 hPa (20 °C)
Refractive Index 1.42
Explosion Limit 13 - 22 %(V)
Density 1.33 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 605 °C DIN 51794
Evaporation number 1.9
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR