Methanol, anhydrous (max. 0.003% H₂O), Seccosolv® analytical reagent, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.06012.1000
1.06012.2500
CA1.06012.1000CS
4036
CAD
CA1.06012.1000
CA1.06012.2500
CA1.06012.6010
Methanol, anhydrous (max. 0.003% H₂O), Seccosolv® analytical reagent, Supelco®
Methanol
These reagents are produced using specially selected distillation methods that ensure consistently high dryness and batch-to-batch consistency. To protect the quality of these products even better from potential contaminants, a septum seal cap provides multiple layers of protection to keep solvents in flawless condition before, during, and after removal.
Formula:
CH₃OH Boiling Pt: 64.6 °C (1013 hPa) Melting Pt: –98 °C Density: 0.7918 g/cm³ (20 °C) Flash Pt: 11 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3 (6.1),II REACH: 01-2119433307-44 Merck Index: 13,05984 |
|
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -98 °C |
Molar Mass | 32.04 g/mol |
Boiling Point | 64.5 °C (1013 hPa) |
Vapor Pressure | 128 hPa (20 °C) |
Flash Point | 10 °C |
Refractive Index | 1.33 |
Explosion Limit | 5.5 - 44 %(V) |
Density | 0.792 g/cm3 (20 °C) |
pH | (H2O) no data available |
Ignition Point | 455 °C DIN 51794 |
Evaporation number | 6.3 |
Water absorption | 1000 g/kg |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...