Methanol, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

1.06002.0500 1.06002.2500
CA1.06002.0500CS 864 CAD
CA1.06002.0500 CA1.06002.2500
Methanol, Uvasol® for spectroscopy, Supelco®
Methanol

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -98 °C
Molar Mass 32.04 g/mol
Boiling Point 64.5 °C (1013 hPa)
Vapor Pressure 128 hPa (20 °C)
Flash Point 10 °C
Refractive Index 1.33
Explosion Limit 5.5 - 44 %(V)
Density 0.792 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 455 °C DIN 51794
Evaporation number 6.3
Water absorption 1000 g/kg
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR