2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

M-Clarity™ quality level = MQ100

1.04718.0500 1.04718.2500
CA1.04718.0500CS 2479 CAD
CA1.04718.0500 CA1.04718.2500
2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®
2,2,4-Trimethylpentane

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99.8 %
Evaporation residue ≤ 0.0002 %
Water ≤ 0.005 %
Color ≤ 10 Hazen
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm) ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm) ≤ 1.0 ppb
Transmission (at 205nm) ≥ 30 %
Transmission (at 215 nm) ≥ 65 %
Transmission (at 220 nm) ≥ 80 %
Transmission (at 225 nm) ≥ 85 %
Transmission (at 235 nm) ≥ 90 %
Transmission (at 245 nm) ≥ 98 %
Transmission (from 255 nm) ≥ 99 %
Absorbance (at 205nm) ≤ 0.52
Absorbance (at 215 nm) ≤ 0.19
Absorbance (at 220 nm) ≤ 0.10
Absorbance (at 225 nm) ≤ 0.07
Absorbance (at 235 nm) ≤ 0.05
Absorbance (at 245 nm) ≤ 0.01
Absorbance (from 255 nm) ≤ 0.004
Meet the requirements of "Specific use spectroscopy" according ACS and Reag. Ph Eur

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR