Hydrazinium sulfate, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hydrazine sulphate salt, Hydrazine sulphate, Hydrazine sulfate, Hydrazine sulfate salt

M-Clarity™ quality level = MQ100

1.04603.0100 1.04603.0500
CA1.04603.0100CS 25824 CAD
CA1.04603.0100 CA1.04603.0500
Hydrazinium sulfate, GR for analysis, Supelco®
Hydrazinium sulphate
Formula: NH₂NH₂·H₂SO₄
MW: 130.12 g/mol
Melting Pt: 254
Density: 1.37 g/cm³ (20 °C)
MDL Number: MFCD00044873
CAS Number: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1 III
Merck Index: 14,04772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=7a677e6f1725cfa1b9bdef05cc04304e6739079e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (iodometric) ≥ 99.0%
Identity passes test
Insoluble matter ≤0.005 %
Chloride (Cl) ≤0.0005 %
Heavy metals (as Pb) ≤0.0005 %
As (Arsenic) ≤ 0.0001 %
Fe (Iron) ≤ 0.001 %
Residue on ignition (as SO.) ≤ 0.05 %
Corresponds to ACS,Reag. Ph Eur


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR