n-Hexane, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

M-Clarity™ quality level = MQ100

1.04372.0500 1.04372.2500
CA1.04372.0500CS 1316 CAD
CA1.04372.0500 CA1.04372.2500
n-Hexane, Uvasol® for spectroscopy, Supelco®
n-Hexane

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99.0 %
Evaporation residue ≤ 0.0002 %
Water ≤ 0.005 %
Color ≤ 10 Hazen
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm) ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm) ≤ 1.0 ppb
Transmission (at 195 nm) ≥ 10 %
Transmission (at 210 nm) ≥ 60 %
Transmission (at 217 nm) ≥ 80 %
Transmission (at 225 nm) ≥ 90 %
Transmission (from 245 nm) ≥ 98 %
Absorbance (at 195 nm) ≤ 1.00
Absorbance (at 210 nm) ≤ 0.22
Absorbance (at 217 nm) ≤ 0.10
Absorbance (at 225 nm) ≤ 0.05
Absorbance (from 245 nm) ≤ 0.01
Meet the requirements of "Specific use spectroscopy" according Reag. Ph Eur


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR