n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

M-Clarity™ quality level = MQ100

1.04371.1000 1.04371.2500 1.04371.4000 1.04371.4000
CA1.04371.1000CS 2196 CAD
CA1.04371.1000 CA1.04371.2500 EM1.04371.4000 CA1.04371.4000
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
n-Hexane

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥98.0 %
Sum of hexane isomers + methylcyclopentane (GC) ≥99.8 %
Identity (IR) conforms
Evaporation residue ≤3.0 mg/l
Water ≤0.01 %
Color ≤ 10 Hazen
GC/ECD (retention range 1,2,4-tri­chlorobenzene to decachlorobiphenyle, individual signals (lindane standard)) ≤ 3 pg/ml
GC/FID (retention range n-undecane to n-tetracontane,individual signals (n-tetradecane standard)) ≤ 3 ng/ml
suitable for residue analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR