m-Cresol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene

M-Clarity™ quality level = MQ200

8.09691.9030 8.09691.0100 8.09691.0500 8.09691.1000 8.09691.2500
CA8.09691.9030EA 1597 CAD
CA8.09691.9030 CA8.09691.0100 CA8.09691.0500 CA8.09691.1000 CA8.09691.2500
m-Cresol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
m-Cresol
Formula: CH₃C₆H₄OH
MW: 108.14 g/mol
Boiling Pt: 202.2
Melting Pt: 11.5
Density: 1.03 g/cm3 (20 °C)
Flash Pt: 73
Storage Temperature: Ambient
MDL Number: MFCD00002302
CAS Number: 108-39-4
UN: 2076
ADR: 6.1,8 II
Merck Index: 13,02604
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce465c84cc8b7c230526b22e6f830300b605d109&vid=72c08ae3da371936f9349196418a240331cb79ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 23.5 g/l (20 °C)
Melting Point 11.5 °C
Molar Mass 108.14 g/mol
Boiling Point 203 °C (1013 hPa)
Vapor Pressure 0.065 hPa (20 °C)
Flash Point 1 %(V)
Refractive Index 86 °C
Density 1.03 g/cm3 (20 °C)
pH 5 (20 g/l, H2O, 20 °C)
Ignition Point 626 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR