Trioctylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.08649.0010 8.08649.0100 8.08649.0500
CA8.08649.0010CS 113401 CAD
CA8.08649.0010 CA8.08649.0100 CA8.08649.0500
Trioctylamine for synthesis, Sigma-Aldrich®
Trioctylamine
Formula: C₂₄H₅₁N
MDL Number: MFCD00009560
CAS Number: 1116-76-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility  - 0.0001 g/l (25 °C)
Melting Point -39 °C
Molar Mass 353.68 g/mol
Boiling Point 365 - 367 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 168 °C
Refractive Index 1.4485 (20 °C, 589 nm)
Density 0.81 g/cm3 (20 °C)
pH 7 (0.4 g/l, H2O, 20 °C)
Ignition Point 315 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR