Pyrrole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1H-Pyrrole

M-Clarity™ quality level = MQ200

8.07492.0010 8.07492.0025 8.07492.0100
CA8.07492.0010CS 7192 CAD
CA8.07492.0010 CA8.07492.0025 CA8.07492.0100
Pyrrole for synthesis, Sigma-Aldrich®
Pyrrole
Formula: C₄H₅N
MW: 67.09 g/mol
Boiling Pt: 130…131 °C (1013 hPa)
Melting Pt: –24 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 39
MDL Number: MFCD00005216
CAS Number: 109-97-7
EINECS: 203-724-7
UN: 1992
ADR: 3 (6.1) III
Merck Index: 12,08198

Order Now

Specification Test Results

Solubility 60 g/l (20 °C)
Melting Point -24 °C
Molar Mass 67.09 g/mol
Boiling Point 130 - 131 °C (1013 hPa)
Vapor Pressure 8.7 hPa (20 °C)
Flash Point 39 °C
Refractive Index 1.5082 (20 °C, 589 nm)
Explosion Limit 3.10 - 14.8 %(V)
Density 0.97 g/cm3 (20 °C)
pH 6 (10 g/l, H2O, 20 °C)
Ignition Point 550 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR