Isopropylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Aminopropane

M-Clarity™ quality level = MQ200

8.07476.0100 8.07476.1000 8.07476.2500
CA8.07476.0100CS 1198 CAD
CA8.07476.0100 CA8.07476.1000 CA8.07476.2500
Isopropylamine for synthesis, Sigma-Aldrich®
Isopropylamine
Formula: C₃H₉N
Boiling Pt: 32.4 °C (1013 hPa)
Melting Pt: –101 °C
Density: 0.69 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008082
CAS Number: 75-31-0
EINECS: 200-860-9
UN: 1221
ADR: 3 (8),I
Merck Index: 13,05229

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Melting Point -101 °C
Molar Mass 59.11 g/mol
Boiling Point 31 - 33 °C (1013 hPa)
Vapor Pressure 632 hPa (20 °C)
Flash Point -37 °C
Refractive Index 1.3746 (20 °C, 589 nm)
Explosion Limit 2 - 10.4 %(V)
Density 0.69 g/cm3 (20 °C)
pH 13 (700 g/l, H2O, 20 °C)
Ignition Point 330 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR