Piperazine, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.07325.0005 8.07325.0250 8.07325.1000 8.07325.9025
CA8.07325.0005CS 67930 CAD
CA8.07325.0005 CA8.07325.0250 CA8.07325.1000 CA8.07325.9025
Piperazine, anhydrous for synthesis, Sigma-Aldrich®
Piperazine
Formula: C₄H₁₀N₂
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: 44…46 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00005953
CAS Number: 110-85-0
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 13,07545

Order Now

Specification Test Results

Solubility 150 g/l (20 °C)
Melting Point 107 - 111 °C
Molar Mass 86.14 g/mol
Bulk Density 400 kg/m3
Vapor Pressure  - 10 hPa (20 °C)
Flash Point 65 °C
Explosion Limit 4 - 14 %(V)
Density 1.1 g/cm3 (20 °C)
pH 12 (150 g/l, H2O, 20 °C)
Ignition Point 320 °C DIN 51794
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR