Methanesulfonic acid zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


MQ200

8.06022.0005 8.06022.0250 8.06022.1000 8.06022.5000 8.06022.9025
CA8.06022.0005CS 6114 CAD
CA8.06022.0005 CA8.06022.0250 CA8.06022.1000 CA8.06022.5000 CA8.06022.9025
Methanesulfonic acid zS, Sigma-Aldrich®
Methanesulphonic acid
Formula: CH₃SO₃H
MW: 96.11 g/mol
Boiling Pt: 217
Melting Pt: 20
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 189 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 2922
ADR: 8 II
Merck Index: 13,05981
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance of substance (visual) colourless to brownish
Assay (acidimetric) ≥ 99.0 %
Density (d 20\u2007°C/ 4\u2007°C) 1.478 - 1.482
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR