Magnesium ≥99.0% (Mg basis), turnings for synthesis, for grignard-reaction, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.05817.0250 8.05817.1000 8.05817.9020 8.05817.9050
CA8.05817.0250CS 932 CAD
CA8.05817.0250 CA8.05817.1000 CA8.05817.9020 CA8.05817.9050
Magnesium ≥99.0% (Mg basis), turnings for synthesis, for grignard-reaction, Sigma-Aldrich®
Magnesium
Formula: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Order Now

Specification Test Results

Solubility (20 °C) (slow decomposition)
Melting Point 651 °C
Molar Mass 24.31 g/mol
Boiling Point 1107 °C
Flash Point 500 °C
Density 1.75 g/cm3 (20 °C)
Ignition Point 450 °C
Hazard Class 4.1
Hazard Class Description Flammable solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR