p-Cresol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.05223.0101 8.05223.0501 8.05223.1001
CA8.05223.0101CS 4746 CAD
CA8.05223.0101 CA8.05223.0501 CA8.05223.1001
p-Cresol for synthesis, Sigma-Aldrich®
p-Cresol
Formula: C₇H₈O
Boiling Pt: 201.8 °C (1013 hPa)
Melting Pt: 32…34 °C
Density: 1.034 g/cm³ (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00002376
CAS Number: 106-44-5
EINECS: 203-398-6
UN: 3455
ADR: 6.1 (8),II
Merck Index: 13,02604

Order Now

Specification Test Results

Solubility 20 g/l (20 °C)
Melting Point 32 - 34 °C
Molar Mass 108.14 g/mol
Boiling Point 202.5 °C (1013 hPa)
Vapor Pressure 0.06 hPa (20 °C)
Flash Point 86 °C
Explosion Limit 1 %(V)
Density 1.034 g/cm3 (20 °C)
Ignition Point 555 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR