1,1,1,3,3,3-Hexafluoro-2-propanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP
M-Clarity™ quality level = MQ200
8.04515.0025
8.04515.0100
8.04515.1000
CA8.04515.0025CS
24686
CAD
CA8.04515.0025
CA8.04515.0100
CA8.04515.1000
1,1,1,3,3,3-Hexafluoro-2-propanol for synthesis, Sigma-Aldrich®
1,1,1,3,3,3-Hexafluoro-2-propanol
Formula:
C₃H₂F₆O MW: 168.04 g/mol Boiling Pt: 59.1 °C (1013 hPa) Melting Pt: –96 °C Density: 1.492 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011651 CAS Number: 920-66-1 EINECS: 213-059-4 UN: 3265 ADR: 8,II Merck Index: 13,09598 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -3 °C |
Molar Mass | 168.03 g/mol |
Boiling Point | 58 °C (1013 hPa) |
Vapor Pressure | 269 hPa (30 °C) |
Flash Point |
|
Refractive Index | 1.2752 (25 °C, 589 nm) |
Density | 1.62 g/cm3 (20 °C) |
pH | 3 - 4 (H2O) Aqueous solution |
Hazard Class | 8 |
Hazard Class Description | Corrosive material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...